Tert-butyl 2-fluoro-5-iodo-3-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

481433-5G 481433-25G 481433-1G 481433-500MG
77629-864EA 1484.23 USD
77629-864 77635-968 77643-280 77647-270
Tert-butyl 2-fluoro-5-iodo-3-methylbenzoate ≥95%
Tert-butyl 2-fluoro-5-iodo-3-methylbenzoate

Precaución: For Research Use Only

Formula: 336.141 g/mol
MW: C₁₂H₁₄FIO₂
Storage Temperature: Ambient
CAS Number: 3031805-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR