2-(2-Fluoro-6-methoxy-4-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

481426-5G 481426-1G 481426-25G 481426-500MG
77629-862EA 1484.23 USD
77629-862 77639-248 77659-708 77651-236
2-(2-Fluoro-6-methoxy-4-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(2-Fluoro-6-methoxy-4-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₄O₂
Storage Temperature: Ambient
CAS Number: 3037704-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR