1-(2-Fluoro-6-methoxy-4-(trifluoromethyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

481425-5G 481425-500MG 481425-25G 481425-1G
77629-860EA 1484.23 USD
77629-860 77652-908 77656-140 77643-278
1-(2-Fluoro-6-methoxy-4-(trifluoromethyl)phenyl)ethanol ≥95%
1-(2-Fluoro-6-methoxy-4-(trifluoromethyl)phenyl)ethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₀F₄O₂
MW: 238.179 g/mol
Storage Temperature: Ambient
CAS Number: 3037692-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR