1-(2,4-Difluoro-3-methylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

481417-5G 481417-1G 481417-25G 481417-500MG
77629-858EA 1484.23 USD
77629-858 77639-244 77656-134 77651-230
1-(2,4-Difluoro-3-methylphenyl)propan-1-ol ≥95%
1-(2,4-Difluoro-3-methylphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 186.198 g/mol
MW: C₁₀H₁₂F₂O
Storage Temperature: Ambient
CAS Number: 3031788-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR