1-(2,4-Difluoro-3-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

481415-5G 481415-25G 481415-1G 481415-500MG
77629-854EA 1484.23 USD
77629-854 77659-706 77643-274 77651-228
1-(2,4-Difluoro-3-methylphenyl)propan-1-one ≥95%
1-(2,4-Difluoro-3-methylphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 184.183 g/mol
MW: C₁₀H₁₀F₂O
Storage Temperature: Ambient
CAS Number: 3031785-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR