3-(2-Chloro-3,6-difluorophenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

481401-1G 481401-5G 481401-500MG
77639-238EA 591 USD
77639-238 77659-698 77647-264
3-(2-Chloro-3,6-difluorophenyl)oxetan-3-ol ≥95%
3-(2-Chloro-3,6-difluorophenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: C₉H₇ClF₂O₂
Storage Temperature: Ambient
CAS Number: 2155441-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR