1-(2-Chloro-3,6-difluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

481400-500MG 481400-5G 481400-1G
77652-904EA 392 USD
77652-904 77659-696 77643-266
1-(2-Chloro-3,6-difluorophenyl)cyclobutanol ≥95%
1-(2-Chloro-3,6-difluorophenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₀H₉ClF₂O
Storage Temperature: Ambient
CAS Number: 2158650-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR