1-(2-Chloro-3,6-difluorophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

481397-5G 481397-1G 481397-500MG
77629-844EA 1484.23 USD
77629-844 77639-236 77647-262
1-(2-Chloro-3,6-difluorophenyl)propan-1-one ≥95%
1-(2-Chloro-3,6-difluorophenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₉H₇ClF₂O
Storage Temperature: Ambient
CAS Number: 2159194-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR