1-(2-Chloro-6-fluoro-3-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

481391-5G 481391-25G 481391-1G 481391-500MG
77629-840EA 1610.92 USD
77629-840 77656-126 77643-258 77651-216
1-(2-Chloro-6-fluoro-3-methylphenyl)cyclobutanol ≥95%
1-(2-Chloro-6-fluoro-3-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂ClFO
Storage Temperature: Ambient
CAS Number: 3031785-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR