(2-Chloro-6-fluoro-3-methylphenyl)(cyclobutyl)methanone ≥95%

Supplier: AOBChem USA

Warning

481389-5G 481389-25G 481389-1G 481389-500MG
77633-366EA 1441 USD
77633-366 77659-688 77643-254 77651-214
(2-Chloro-6-fluoro-3-methylphenyl)(cyclobutyl)methanone ≥95%
(2-Chloro-6-fluoro-3-methylphenyl)(cyclobutyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₂ClFO
Storage Temperature: Ambient
CAS Number: 3031804-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR