(2-Chloro-6-fluoro-3-methylphenyl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

481388-1G 481388-5G 481388-500MG
77639-232EA 531 USD
77639-232 77656-124 77647-260
(2-Chloro-6-fluoro-3-methylphenyl)(cyclopropyl)methanone ≥95%
(2-Chloro-6-fluoro-3-methylphenyl)(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: 212.648 g/mol
MW: C₁₁H₁₀ClFO
Storage Temperature: Ambient
CAS Number: 1534171-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR