1-(2,3-Dichloro-6-fluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

481384-5G 481384-1G 481384-25G 481384-500MG
77629-836EA 1484.23 USD
77629-836 77639-230 77656-122 77647-258
1-(2,3-Dichloro-6-fluorophenyl)cyclobutanol ≥95%
1-(2,3-Dichloro-6-fluorophenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 235.082 g/mol
MW: C₁₀H₉Cl₂FO
Storage Temperature: Ambient
CAS Number: 3031812-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR