1,2,3-Trichloro-4-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

481383-5G 481383-1G 481383-25G 481383-500MG
77633-364EA 833 USD
77633-364 77639-228 77659-686 77651-212
1,2,3-Trichloro-4-fluorobenzene ≥95%
1,2,3-Trichloro-4-fluorobenzene

Precaución: For Research Use Only

Formula: C₆H₂Cl₃F
Storage Temperature: Ambient
CAS Number: 36556-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR