Ethyl 2-(2,3-dichloro-6-fluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

481381-5G 481381-25G 481381-1G 481381-500MG
77629-834EA 1484.23 USD
77629-834 77635-964 77639-226 77647-256
Ethyl 2-(2,3-dichloro-6-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,3-dichloro-6-fluorophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 265.065 g/mol
MW: C₁₀H₇Cl₂FO₃
Storage Temperature: Ambient
CAS Number: 3031789-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR