Cyclopropyl(2,3-dichloro-6-fluorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

481380-5G 481380-25G 481380-1G 481380-500MG
77629-832EA 1484.23 USD
77629-832 77635-962 77639-224 77647-254
Cyclopropyl(2,3-dichloro-6-fluorophenyl)methanone ≥95%
Cyclopropyl(2,3-dichloro-6-fluorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₇Cl₂FO
Storage Temperature: Ambient
CAS Number: 3031789-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR