1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

481374-5G 481374-25G 481374-1G 481374-500MG
77629-828EA 1484.23 USD
77629-828 77656-118 77643-250 77651-210
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)cyclobutanol ≥95%
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₉F₅O
Storage Temperature: Ambient
CAS Number: 3031789-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR