Ethyl 2-(2,3-difluoro-4-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

481372-5G 481372-25G 481372-1G 481372-500MG
77629-826EA 1484.23 USD
77629-826 77635-960 77643-248 77647-250
Ethyl 2-(2,3-difluoro-4-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,3-difluoro-4-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₁H₇F₅O₃
Storage Temperature: Ambient
CAS Number: 3031798-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR