1-(4-Bromobenzo[b]thiophen-2-yl)-2-chloroethanone ≥95%

Supplier: AOBChem USA

Warning

481363-5G 481363-1G 481363-25G 481363-500MG
77629-822EA 1505.86 USD
77629-822 77639-218 77656-116 77651-206
1-(4-Bromobenzo[b]thiophen-2-yl)-2-chloroethanone ≥95%
1-(4-Bromobenzo[b]thiophen-2-yl)-2-chloroethanone

Precaución: For Research Use Only

Formula: C₁₀H₆BrClOS
Storage Temperature: Ambient
CAS Number: 3031804-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR