2,4-Dimethoxy-1-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

481264-5G 481264-1G 481264-25G 481264-500MG
77633-360EA 1135 USD
77633-360 77639-216 77659-680 77647-246
2,4-Dimethoxy-1-(methoxymethoxy)benzene ≥97%
2,4-Dimethoxy-1-(methoxymethoxy)benzene

Precaución: For Research Use Only

Formula: 198.216 g/mol
MW: C₁₀H₁₄O₄
Storage Temperature: Ambient
CAS Number: 104202-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR