5-Bromo-3-ethyl-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

480849-5G 480849-25G 480849-1G 480849-500MG
77633-358EA 898 USD
77633-358 77656-110 77643-244 77651-202
5-Bromo-3-ethyl-1H-indazole ≥95%
5-Bromo-3-ethyl-1H-indazole

Precaución: For Research Use Only

Formula: C₉H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD11044592
CAS Number: 864774-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR