N-Methoxy-N,3-dimethylbut-2-enamide ≥97%

Supplier: AOBChem USA

Warning

480637-5G 480637-1G 480637-25G 480637-500MG
77633-356EA 1328 USD
77633-356 77639-214 77656-108 77651-200
N-Methoxy-N,3-dimethylbut-2-enamide ≥97%
N-Methoxy-N,3-dimethylbut-2-enamide

Precaución: For Research Use Only

Formula: 143.1836 g/mol
MW: C₇H₁₃NO₂
Storage Temperature: Ambient
CAS Number: 121712-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR