2,6-Diiodo-4-(trifluoromethoxy)phenol ≥97%

Supplier: AOBChem USA

Warning

480596-5G 480596-1G 480596-25G 480596-500MG
77629-818EA 1484.23 USD
77629-818 77639-212 77659-678 77651-198
2,6-Diiodo-4-(trifluoromethoxy)phenol ≥97%
2,6-Diiodo-4-(trifluoromethoxy)phenol

Precaución: For Research Use Only

Formula: C₇H₃F₃I₂O₂
MW: 429.9016 g/mol
Storage Temperature: Ambient
CAS Number: 405517-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR