(2,3-Difluoro-4-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

480574-5G 480574-25G 480574-1G 480574-500MG
77633-354EA 1441 USD
77633-354 77659-676 77643-242 77651-196
(2,3-Difluoro-4-methylphenyl)(phenyl)methanol ≥95%
(2,3-Difluoro-4-methylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₄H₁₂F₂O
Storage Temperature: Ambient
CAS Number: 1930998-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR