3-(4-Chloro-2,3-difluorophenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

480572-5G 480572-25G 480572-1G 480572-500MG
77629-816EA 1484.23 USD
77629-816 77659-674 77643-240 77647-244
3-(4-Chloro-2,3-difluorophenyl)oxetan-3-ol ≥95%
3-(4-Chloro-2,3-difluorophenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: 220.6 g/mol
MW: C₉H₇ClF₂O₂
Storage Temperature: Ambient
CAS Number: 3031804-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR