(4-Chloro-2,3-difluorophenyl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

480571-5G 480571-25G 480571-1G 480571-500MG
77633-352EA 1441 USD
77633-352 77656-106 77643-238 77651-194
(4-Chloro-2,3-difluorophenyl)(cyclopropyl)methanone ≥95%
(4-Chloro-2,3-difluorophenyl)(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: 216.612 g/mol
MW: C₁₀H₇ClF₂O
Storage Temperature: Ambient
CAS Number: 3031785-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR