1-(4-Chloro-2,3-difluorophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

480570-5G 480570-1G 480570-500MG 480570-25G
77629-814EA 1484.23 USD
77629-814 77639-210 77652-898 77656-104
1-(4-Chloro-2,3-difluorophenyl)propan-1-one ≥95%
1-(4-Chloro-2,3-difluorophenyl)propan-1-one

Precaución: For Research Use Only

Formula: 204.601 g/mol
MW: C₉H₇ClF₂O
Storage Temperature: Ambient
CAS Number: 3031789-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR