Methyl 2-(4-chloro-2,3-difluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

480569-5G 480569-25G 480569-1G 480569-500MG
77629-812EA 1627.4 USD
77629-812 77635-958 77643-236 77651-192
Methyl 2-(4-chloro-2,3-difluorophenyl)-2-oxoacetate ≥95%
Methyl 2-(4-chloro-2,3-difluorophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₉H₅ClF₂O₃
Storage Temperature: Ambient
CAS Number: 3031784-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR