Cyclopropyl(3,4-dichloro-2-fluorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

480566-5G 480566-25G 480566-500MG 480566-1G
77629-810EA 1484.23 USD
77629-810 77635-956 77652-896 77643-232
Cyclopropyl(3,4-dichloro-2-fluorophenyl)methanone ≥95%
Cyclopropyl(3,4-dichloro-2-fluorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₇Cl₂FO
Storage Temperature: Ambient
CAS Number: 3031789-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR