1-(3,4-Dichloro-2-fluorophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

480565-5G 480565-25G 480565-1G 480565-500MG
77629-808EA 1601.65 USD
77629-808 77656-100 77643-230 77647-240
1-(3,4-Dichloro-2-fluorophenyl)propan-1-one ≥95%
1-(3,4-Dichloro-2-fluorophenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₉H₇Cl₂FO
Storage Temperature: Ambient
CAS Number: 3031789-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR