3-(3,4-Dichloro-2-fluorophenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

480564-5G 480564-1G 480564-25G 480564-500MG
77629-806EA 1484.23 USD
77629-806 77639-208 77659-672 77647-238
3-(3,4-Dichloro-2-fluorophenyl)oxetan-3-ol ≥95%
3-(3,4-Dichloro-2-fluorophenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: C₉H₇Cl₂FO₂
Storage Temperature: Ambient
CAS Number: 3031798-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR