Ethyl 2-(3,4-dichloro-2-fluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

480561-5G 480561-1G 480561-500MG
77629-804EA 1484.23 USD
77629-804 77639-206 77647-236
Ethyl 2-(3,4-dichloro-2-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(3,4-dichloro-2-fluorophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₀H₇Cl₂FO₃
Storage Temperature: Ambient
CAS Number: 3037682-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR