3-(2,3-Difluoropyridin-4-yl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

480559-5G 480559-25G 480559-1G 480559-500MG
77629-802EA 1484.23 USD
77629-802 77656-098 77643-228 77647-234
3-(2,3-Difluoropyridin-4-yl)oxetan-3-ol ≥95%
3-(2,3-Difluoropyridin-4-yl)oxetan-3-ol

Precaución: For Research Use Only

Formula: 187.143 g/mol
MW: C₈H₇F₂NO₂
Storage Temperature: Ambient
CAS Number: 3037705-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR