1-(2,3-Difluoropyridin-4-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

480557-5G 480557-25G 480557-1G 480557-500MG
77629-800EA 1484.23 USD
77629-800 77659-670 77643-226 77647-232
1-(2,3-Difluoropyridin-4-yl)propan-1-one ≥95%
1-(2,3-Difluoropyridin-4-yl)propan-1-one

Precaución: For Research Use Only

Formula: 171.144 g/mol
MW: C₈H₇F₂NO
Storage Temperature: Ambient
CAS Number: 3037676-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR