1-(3-Fluoro-2-methoxypyridin-4-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

480552-1G 480552-500MG 480552-5G
77639-202EA 676 USD
77639-202 77652-894 77656-096
1-(3-Fluoro-2-methoxypyridin-4-yl)cyclobutanol ≥95%
1-(3-Fluoro-2-methoxypyridin-4-yl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₀H₁₂FNO₂
Storage Temperature: Ambient
CAS Number: 1850191-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR