Cyclopropyl(3-fluoro-2-methoxypyridin-4-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

480550-1G 480550-5G 480550-500MG
77639-198EA 628 USD
77639-198 77659-666 77647-228
Cyclopropyl(3-fluoro-2-methoxypyridin-4-yl)methanone ≥95%
Cyclopropyl(3-fluoro-2-methoxypyridin-4-yl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₁₀FNO₂
Storage Temperature: Ambient
CAS Number: 1880683-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR