1-(3-Fluoro-2-methoxypyridin-4-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

480549-500MG 480549-5G 480549-1G
77652-892EA 379 USD
77652-892 77659-664 77643-224
1-(3-Fluoro-2-methoxypyridin-4-yl)propan-1-one ≥95%
1-(3-Fluoro-2-methoxypyridin-4-yl)propan-1-one

Precaución: For Research Use Only

Formula: 183.18 g/mol
MW: C₉H₁₀FNO₂
Storage Temperature: Ambient
CAS Number: 1864176-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR