Ethyl 2-(5-bromobenzofuran-2-yl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

480544-5G 480544-25G 480544-1G 480544-500MG
77629-796EA 1484.23 USD
77629-796 77635-952 77639-196 77652-890
Ethyl 2-(5-bromobenzofuran-2-yl)-2-oxoacetate ≥95%
Ethyl 2-(5-bromobenzofuran-2-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 297.101 g/mol
MW: C₁₂H₉BrO₄
Storage Temperature: Ambient
CAS Number: 1506492-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR