Cyclohexyl(2,5-dichlorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

480538-5G 480538-1G 480538-500MG
77629-794EA 1484.23 USD
77629-794 77643-222 77651-188
Cyclohexyl(2,5-dichlorophenyl)methanone ≥97%
Cyclohexyl(2,5-dichlorophenyl)methanone

Precaución: For Research Use Only

Formula: 257.156 g/mol
MW: C₁₃H₁₄Cl₂O
Storage Temperature: Ambient
CAS Number: 898769-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR