1-(2,5-Dichlorophenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

480536-5G 480536-1G 480536-500MG
77629-792EA 1484.23 USD
77629-792 77639-192 77647-224
1-(2,5-Dichlorophenyl)-2-fluoroethanone ≥95%
Formula: C₈H₅Cl₂FO
Storage Temperature: Ambient
CAS Number: 2294736-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR