(2,5-Dichlorophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

480477-5G 480477-25G 480477-1G 480477-500MG
77633-348EA 1308 USD
77633-348 77659-662 77643-220 77651-186
(2,5-Dichlorophenyl)(phenyl)methanol ≥95%
(2,5-Dichlorophenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 253.124 g/mol
MW: C₁₃H₁₀Cl₂O
Storage Temperature: Ambient
CAS Number: 1094791-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR