2-(2,5-Dichlorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

480471-5G 480471-1G 480471-25G 480471-500MG
77633-346EA 1326 USD
77633-346 77639-190 77659-660 77647-222
2-(2,5-Dichlorophenyl)propan-2-ol ≥95%
2-(2,5-Dichlorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 205.081 g/mol
MW: C₉H₁₀Cl₂O
Storage Temperature: Ambient
CAS Number: 62436-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR