1-(2,6-Difluoro-4-methoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

480245-5G 480245-1G 480245-500MG
77629-788EA 1484.23 USD
77629-788 77639-186 77651-180
1-(2,6-Difluoro-4-methoxyphenyl)propan-1-one ≥95%
1-(2,6-Difluoro-4-methoxyphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 200.1820 g/mol
MW: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
CAS Number: 1183525-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR