Ethyl 7-bromo-3-cyclopropylbenzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

480236-5G 480236-1G 480236-500MG
77629-786EA 1484.23 USD
77629-786 77643-218 77651-178
Ethyl 7-bromo-3-cyclopropylbenzo[b]thiophene-2-carboxylate ≥95%
Ethyl 7-bromo-3-cyclopropylbenzo[b]thiophene-2-carboxylate

Precaución: For Research Use Only

Formula: 325.2208 g/mol
MW: C₁₄H₁₃BrO₂S
Storage Temperature: Ambient
CAS Number: 1517557-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR