1-Benzyl-5-bromo-1H-imidazole ≥97%

Supplier: AOBChem USA

Warning

480086-5G 480086-500MG 480086-25G 480086-1G
77633-344EA 727 USD
77633-344 77652-888 77656-090 77643-216
1-Benzyl-5-bromo-1H-imidazole ≥97%
1-Benzyl-5-bromo-1H-imidazole

Precaución: For Research Use Only

Formula: C₁₀H₉BrN₂
MW: 237.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19688638
CAS Number: 132430-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR