Methyl 3-amino-5-iodo-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

480080-5G 480080-25G 480080-1G 480080-500MG
77629-780EA 1484.23 USD
77629-780 77635-950 77639-180 77647-216
Methyl 3-amino-5-iodo-4-methylbenzoate ≥97%
Methyl 3-amino-5-iodo-4-methylbenzoate

Precaución: For Research Use Only

Formula: C₉H₁₀INO₂
Storage Temperature: Ambient
CAS Number: 21324-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR