1-[4-(Dimethylamino)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

480016-5G 480016-1G 480016-25G 480016-500MG
77633-342EA 712 USD
77633-342 77639-178 77656-088 77651-174
1-[4-(Dimethylamino)phenyl]-1-propanone ≥95%
1-[4-(Dimethylamino)phenyl]-1-propanone

Precaución: For Research Use Only

Formula: 177.24 g/mol
MW: C₁₁H₁₅NO
Storage Temperature: Ambient
CAS Number: 26672-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR