2-Bromo-4-chloro-6-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47904-5G 47904-1G 47904-25G 47904-500MG
77629-778EA 1484.23 USD
77629-778 77639-176 77656-084 77647-212
2-Bromo-4-chloro-6-fluoro-3-methylbenzaldehyde ≥97%
2-Bromo-4-chloro-6-fluoro-3-methylbenzaldehyde

Precaución: For Research Use Only

Formula: 251.48 g/mol
MW: C₈H₅BrClFO
Storage Temperature: Ambient
CAS Number: 2368909-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR