2,2,2-Trifluoro-1-(2-isopropyl-4-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

478482-5G 478482-1G 478482-500MG
77629-776EA 1484.23 USD
77629-776 77639-174 77647-210
2,2,2-Trifluoro-1-(2-isopropyl-4-methoxyphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-isopropyl-4-methoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 246.226 g/mol
MW: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
CAS Number: 3037705-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR