1-(2,5-Diethylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

478028-5G 478028-1G 478028-25G 478028-500MG
77629-766EA 1484.23 USD
77629-766 77639-164 77659-652 77647-200
1-(2,5-Diethylphenyl)cyclobutanol ≥95%
1-(2,5-Diethylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 204.308 g/mol
MW: C₁₄H₂₀O
Storage Temperature: Ambient
CAS Number: 3037682-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR