Tert-butyl 2,5-diethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

478025-5G 478025-25G 478025-1G 478025-500MG
77629-764EA 1484.23 USD
77629-764 77635-944 77643-206 77651-164
Tert-butyl 2,5-diethylbenzoate ≥95%
Tert-butyl 2,5-diethylbenzoate

Precaución: For Research Use Only

Formula: 234.334 g/mol
MW: C₁₅H₂₂O₂
Storage Temperature: Ambient
CAS Number: 3037696-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR